Assay Details
Gene Symbol: Fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158967.1 | - | 317 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK158967.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
