Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100019778.mm.1 |
NM_001177874.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019778.mm.1 |
NM_001177874.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019778.mm.1 |
NM_145370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019778.mm.1 |
NM_145370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019778.mm.1 |
TC1100002048.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100019779.mm.1 |
NM_001177874.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019779.mm.1 |
NM_001177874.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019779.mm.1 |
NM_145370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019779.mm.1 |
NM_145370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100019779.mm.1 |
TC1100002048.mm.1 |
MTA TC |