Assay Details
| Gene Symbol: Cd300e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172050.2 | NP_742047.1 | 3 - 4 | 507 | 83 | |
| GenBank mRNA | AF437879.1 | - | 3 - 4 | 531 | 83 | |
| AY457048.1 | - | 3 - 4 | 493 | 83 | ||
| BC034097.1 | - | 3 - 4 | 532 | 4487018 | 83 | |
| BC039971.1 | - | 3 - 4 | 543 | 4487018 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312035 | BC034097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312035 | BC039971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312035 | NM_172050.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312035 | BC034097.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312035 | BC039971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P312035 | NM_172050.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94169 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178326 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104838 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312035 | BC034097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312035 | BC039971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312035 | NM_172050.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312035 | BC034097.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312035 | BC039971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P312035 | NM_172050.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28151171 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104838 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178326 | - | - |
| Whole Mouse Genome G4122A | A_51_P312035 | BC034097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312035 | BC039971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312035 | NM_172050.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312035 | BC034097.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312035 | BC039971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P312035 | NM_172050.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128643 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104838 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178326 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09631871 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09631861 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09631863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28151206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09391600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09476732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09391604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09726726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09717625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09617114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09631868 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09443876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09580767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09612515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09447251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09399032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09259320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09363854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28151160 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10392842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10392843 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10392839 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100038072.mm.1 | NM_172050.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038072.mm.1 | TC1100004059.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100038071.mm.1 | NM_172050.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100038071.mm.1 | TC1100004059.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 808161 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2120041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2120041 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002120041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1212716 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1212716 | - | - |
| mouse-6 v1.1 (Illumina) | scl39349.4.1_146-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2120041 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39349.4.1_146-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2120041 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1212716 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1212716 | - | - |

Cd300e
Genomic Map