Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100038275.mm.1 |
NM_015807.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038275.mm.1 |
NM_015807.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038275.mm.1 |
TC1100004075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100038278.mm.1 |
NM_015807.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038278.mm.1 |
NM_015807.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038278.mm.1 |
TC1100004075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100038277.mm.1 |
NM_015807.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038277.mm.1 |
NM_015807.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038277.mm.1 |
TC1100004075.mm.1 |
MTA TC |