Assay Details
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CK619967.1 | - | 3 - 4 | 175 | 106 | |
DQ092438.1 | - | 2 - 3 | 61 | 106 | ||
DQ251193.1 | - | 2 - 3 | 61 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the DQ251193.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
