Assay Details
| Gene Symbol: Gcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008101.2 | NP_032127.2 | 10 - 11 | 1132 | 76 | |
| GenBank mRNA | AK004890.1 | - | 7 - 8 | 1339 | 76 | |
| AK144393.1 | - | 10 - 11 | 1079 | 76 | ||
| BC012718.1 | - | 9 - 10 | 1207 | 76 | ||
| BC031885.1 | - | 10 - 11 | 1082 | 5137340 | 76 | |
| BC057988.1 | - | 10 - 11 | 1063 | 5100574 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gcgr
Genomic Map