Assay Details
| Gene Symbol: Gcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008101.2 | NP_032127.2 | 11 - 12 | 1225 | 75 | |
| GenBank mRNA | AK004890.1 | - | 8 - 9 | 1432 | 75 | |
| AK144393.1 | - | 11 - 12 | 1172 | 75 | ||
| BC012718.1 | - | 10 - 11 | 1300 | 75 | ||
| BC031885.1 | - | 11 - 12 | 1175 | 5137340 | 75 | |
| BC057988.1 | - | 11 - 12 | 1156 | 5100574 | 75 | |
| S69384.1 | - | 1 - 2 | 72 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gcgr
Genomic Map