Assay Details
| Gene Symbol: Slc26a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178743.3 | NP_848858.2 | 13 - 14 | 1495 | 72 | |
| GenBank mRNA | AF345196.1 | - | 13 - 14 | 1495 | 72 | |
| AK052202.1 | - | 8 - 9 | 1582 | 72 | ||
| AK089182.1 | - | 13 - 14 | 1475 | 72 | ||
| BC038604.1 | - | 13 - 14 | 1488 | 72 | ||
| BC132493.1 | - | 8 - 9 | 1363 | 40130770 | 72 | |
| BC137897.1 | - | 8 - 9 | 1363 | 8860916 | 72 | |
More Information
Set Membership: |
|

Slc26a11
Genomic Map