Assay Details
| Gene Symbol: Slc26a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178743.3 | NP_848858.2 | 2 - 3 | 304 | 68 | |
| GenBank mRNA | AF345196.1 | - | 2 - 3 | 304 | 68 | |
| AK078545.1 | - | 2 - 3 | 280 | 68 | ||
| AK089182.1 | - | 2 - 3 | 284 | 68 | ||
| BC038604.1 | - | 2 - 3 | 297 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a11
Genomic Map