Assay Details
| Gene Symbol: Sept9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113486.1 | NP_001106958.1 | 1 - 2 | 115 | 112 | |
| GenBank mRNA | AK141312.1 | - | 1 - 2 | 56 | 112 | |
| BC046524.1 | - | 1 - 2 | 115 | 5290621 | 112 | |
| BC094455.1 | - | 1 - 2 | 38 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK141312.1, BC046524.1, NM_001113486.1 transcripts.
Set Membership: |
|

Sept9
Genomic Map