Assay Details
| Gene Symbol: Pgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133757.2 | NP_598518.1 | 7 - 8 | 1421 | 119 | |
| GenBank mRNA | AK046174.1 | - | 7 - 8 | 1413 | 119 | |
| AK049359.1 | - | 7 - 8 | 1422 | 119 | ||
| AK053905.1 | - | 7 - 8 | 1407 | 119 | ||
| AK153001.1 | - | 5 - 6 | 1114 | 119 | ||
| AK178063.1 | - | 1 - 2 | 274 | 119 | ||
| AK181137.1 | - | 1 - 2 | 118 | 119 | ||
| BC022918.1 | - | 2 - 3 | 543 | 119 | ||
| BC151089.1 | - | 7 - 8 | 1521 | 9088002 | 119 | |
| BC151185.1 | - | 7 - 8 | 1521 | 9088074 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgs1
Genomic Map