Assay Details
Gene Symbol: Mgat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172948.3 | NP_766536.2 | 3 - 4 | 944 | 60 | |
GenBank mRNA | AB116028.1 | - | 3 - 4 | 331 | 60 | |
AB119127.1 | - | 3 - 4 | 410 | 60 | ||
BC063084.1 | - | 3 - 4 | 944 | 60 | ||
BC094604.1 | - | 3 - 4 | 944 | 6822690 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
