Assay Details
Gene Symbol: Cbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007623.2 | NP_031649.2 | 3 - 4 | 268 | 61 | |
GenBank mRNA | BC035199.1 | - | 3 - 4 | 268 | 5137505 | 61 |
X62537.1 | - | 3 - 4 | 335 | 61 | ||
Y13274.1 | - | 3 - 4 | 192 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
