Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800009339.mm.1 |
NM_001079865.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009339.mm.1 |
NM_001079865.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009339.mm.1 |
TC0800001197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009341.mm.1 |
NM_001079865.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009341.mm.1 |
NM_001079865.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009341.mm.1 |
TC0800001197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009338.mm.1 |
NM_001079865.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009338.mm.1 |
NM_001079865.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009338.mm.1 |
TC0800001197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009340.mm.1 |
NM_001079865.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009340.mm.1 |
NM_001079865.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009340.mm.1 |
TC0800001197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009342.mm.1 |
NM_001079865.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009342.mm.1 |
NM_001079865.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009342.mm.1 |
TC0800001197.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009343.mm.1 |
NM_001079865.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009343.mm.1 |
NM_001079865.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009343.mm.1 |
TC0800001197.mm.1 |
MTA TC |