Assay Details
| Gene Symbol: Ces2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079865.2 | NP_001073334.2 | 6 - 7 | 937 | 65 | |
| GenBank mRNA | AK040349.1 | - | 6 - 7 | 939 | 65 | |
| BC117742.1 | - | 6 - 7 | 937 | 40090393 | 65 | |
| BC117743.1 | - | 4 - 5 | 544 | 40090395 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ces2f
Genomic Map