Assay Details
| Gene Symbol: Fendrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087264.1 | - | 3 - 4 | 799 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 26878 |
Gm26878 | -183.6kb | Anti-sense | Chr. 8 - 120879399 - 120881217 on NCBI Build GRCm38 | |
| Downstream | forkhead box F1 | Foxf1 |
AI450827, FREAC1, Foxf1a, Freac-1, HFH-8, Hfh8 |
18.6kb | Anti-sense | Chr. 8 - 121084386 - 121088154 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Fendrr
Genomic Map