Assay Details
| Gene Symbol: Ces1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144930.2 | NP_659179.1 | 7 - 8 | 939 | 92 | |
| GenBank mRNA | AB023632.1 | - | 3 - 4 | 342 | 92 | |
| AI786051.1 | - | 3 - 4 | 335 | 92 | ||
| AK136491.1 | - | 7 - 8 | 931 | 92 | ||
| BC013479.1 | - | 7 - 8 | 914 | 4239756 | 92 | |
| EH107325.1 | - | 2 - 3 | 187 | 92 | ||
More Information
Set Membership: |
|

Ces1f
Genomic Map