Assay Details
| Gene Symbol: Ces1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053200.2 | NP_444430.2 | 8 - 9 | 1022 | 103 | |
| GenBank mRNA | AB023631.1 | - | 8 - 9 | 982 | 103 | |
| AF378751.1 | - | 8 - 9 | 997 | 103 | ||
| AK078879.1 | - | 8 - 9 | 1000 | 103 | ||
| BC019198.1 | - | 8 - 9 | 994 | 5040523 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ces1d
Genomic Map