| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800010075.mm.1 |
NM_001037809.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800010075.mm.1 |
NM_007665.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800010075.mm.1 |
TC0800001253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800010079.mm.1 |
NM_001037809.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800010079.mm.1 |
NM_007665.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800010079.mm.1 |
TC0800001253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800010078.mm.1 |
NM_001037809.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800010078.mm.1 |
NM_007665.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800010078.mm.1 |
TC0800001253.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800010077.mm.1 |
NM_001037809.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800010077.mm.1 |
NM_007665.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800010077.mm.1 |
TC0800001253.mm.1 |
MTA TC |