Assay Details
| Gene Symbol: Cdh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037809.5 | NP_001032898.1 | 5 - 6 | 621 | 71 | |
| NM_007665.3 | NP_031691.1 | 5 - 6 | 618 | 71 | ||
| GenBank mRNA | AK031265.1 | - | 5 - 6 | 561 | 71 | |
| AK045041.1 | - | 5 - 6 | 581 | 71 | ||
| AK164577.1 | - | 5 - 6 | 591 | 71 | ||
| BC052189.1 | - | 3 - 4 | 309 | 71 | ||
| BC098459.1 | - | 5 - 6 | 527 | 30079668 | 71 | |
| X06340.1 | - | 5 - 6 | 559 | 71 | ||
More Information
Set Membership: |
|

Cdh3
Genomic Map