Assay Details
| Gene Symbol: Pgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133757.2 | NP_598518.1 | 4 - 5 | 529 | 106 | |
| GenBank mRNA | AK016901.1 | - | 2 - 3 | 348 | 106 | |
| AK046174.1 | - | 4 - 5 | 521 | 106 | ||
| AK049359.1 | - | 4 - 5 | 530 | 106 | ||
| AK053905.1 | - | 4 - 5 | 515 | 106 | ||
| AK153001.1 | - | 2 - 3 | 222 | 106 | ||
| AK163967.1 | - | 4 - 5 | 502 | 106 | ||
| AK189295.1 | - | 3 - 4 | 279 | 106 | ||
| BC151089.1 | - | 4 - 5 | 629 | 9088002 | 106 | |
| BC151185.1 | - | 4 - 5 | 629 | 9088074 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgs1
Genomic Map