Assay Details
Gene Symbol: Pgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133757.2 | NP_598518.1 | 6 - 7 | 901 | 72 | |
GenBank mRNA | AK016901.1 | - | 4 - 5 | 720 | 72 | |
AK046174.1 | - | 6 - 7 | 893 | 72 | ||
AK049359.1 | - | 6 - 7 | 902 | 72 | ||
AK053905.1 | - | 6 - 7 | 887 | 72 | ||
AK153001.1 | - | 4 - 5 | 594 | 72 | ||
AK163967.1 | - | 6 - 7 | 875 | 72 | ||
BC151089.1 | - | 6 - 7 | 1001 | 9088002 | 72 | |
BC151185.1 | - | 6 - 7 | 1001 | 9088074 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
