Assay Details
| Gene Symbol: Spns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276383.1 | NP_001263312.1 | 11 - 12 | 1382 | 92 | |
| NM_153060.3 | NP_694700.2 | 11 - 12 | 1730 | 92 | ||
| GenBank mRNA | AK156007.1 | - | 12 - 13 | 1376 | 92 | |
| BC011467.1 | - | 11 - 12 | 1730 | 92 | ||
| BC025823.1 | - | 2 - 3 | 404 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spns2
Genomic Map