Assay Details
| Gene Symbol: Spns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153060.3 | NP_694700.2 | 1 - 2 | 495 | 71 | |
| GenBank mRNA | AB441166.1 | - | 1 - 2 | 367 | 71 | |
| AK156007.1 | - | 2 - 3 | 141 | 71 | ||
| BC011467.1 | - | 1 - 2 | 495 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spns2
Genomic Map