Assay Details
| Gene Symbol: Spns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276383.1 | NP_001263312.1 | 3 - 4 | 349 | 64 | |
| NM_153060.3 | NP_694700.2 | 3 - 4 | 697 | 64 | ||
| GenBank mRNA | AB441166.1 | - | 3 - 4 | 569 | 64 | |
| AK156007.1 | - | 4 - 5 | 343 | 64 | ||
| BC011467.1 | - | 3 - 4 | 697 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spns2
Genomic Map