Assay Details
| Gene Symbol: Spns2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276383.1 | NP_001263312.1 | 6 - 7 | 715 | 55 | |
| NM_153060.3 | NP_694700.2 | 6 - 7 | 1063 | 55 | ||
| GenBank mRNA | AB441166.1 | - | 6 - 7 | 935 | 55 | |
| AK156007.1 | - | 7 - 8 | 709 | 55 | ||
| BC011467.1 | - | 6 - 7 | 1063 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spns2
Genomic Map