Assay Details
| Gene Symbol: Mrpl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027204.2 | NP_081480.2 | 3 - 4 | 379 | 81 | |
| GenBank mRNA | AK002757.1 | - | 3 - 4 | 389 | 81 | |
| AK005328.1 | - | 3 - 4 | 410 | 81 | ||
| AK028168.1 | - | 3 - 4 | 390 | 81 | ||
| AK134816.1 | - | 3 - 4 | 412 | 81 | ||
| AK166969.1 | - | 3 - 4 | 408 | 81 | ||
| AK168337.1 | - | 3 - 4 | 382 | 81 | ||
| BC005712.1 | - | 3 - 4 | 379 | 3582724 | 81 | |
| BC019565.1 | - | 3 - 4 | 377 | 4219731 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28184854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28184855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28184856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28184858 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | AK166969.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | AK168337.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | AK134816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | BC005712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | BC019565.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | AK166969.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | AK168337.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | AK134816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | BC005712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143142 | BC019565.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153027 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135293 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | AK166969.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | AK168337.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | AK134816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | BC005712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | BC019565.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | AK166969.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | AK168337.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | AK134816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | BC005712.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143142 | BC019565.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | AK166969.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | AK168337.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | AK134816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | BC005712.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | BC019565.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | AK166969.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | AK168337.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | AK134816.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | BC005712.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143142 | BC019565.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184854 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135293 | - | - |
| Whole Mouse Genome G4122A | A_51_P143142 | AK166969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | AK168337.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | AK134816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | BC005712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | BC019565.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | AK166969.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | AK168337.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | AK134816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | BC005712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143142 | BC019565.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140685 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117773 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135293 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09186861 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09186891 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09186888 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09186876 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28184856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28184854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28184855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28184858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09585948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184870 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09585949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09244960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09186889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28184860 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10535371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383393 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452048_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10535369 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10383389 | - | - |
| Mouse Genome 430 2.0 Array | 1452048_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452048_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56282_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56282_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100019537.mm.1 | NM_027204.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019537.mm.1 | NM_027204.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019537.mm.1 | TC1100002029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019536.mm.1 | NM_027204.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019536.mm.1 | NM_027204.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019536.mm.1 | TC1100002029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019539.mm.1 | NM_027204.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019539.mm.1 | NM_027204.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019539.mm.1 | TC1100002029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019540.mm.1 | NM_027204.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019540.mm.1 | NM_027204.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019540.mm.1 | TC1100002029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019538.mm.1 | NM_027204.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019538.mm.1 | NM_027204.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019538.mm.1 | TC1100002029.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100019534.mm.1 | NM_027204.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019534.mm.1 | NM_027204.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100019534.mm.1 | TC1100002029.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 160431_at | - | - |
| Murine Genome U74v2 Set | 160431_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452048_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452048_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 630492 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3120112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003120112 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2735335 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2735335 | - | - |
| mouse-6 v1.1 (Illumina) | scl056282.5_222-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3120112 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl056282.5_222-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3120112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2883147 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2883147 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl056282.5_222-S | - | - |

Mrpl12
Genomic Map