Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1430740_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430740_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430740_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1100036531.mm.1 |
NR_104047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036531.mm.1 |
TC1100003909.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036526.mm.1 |
NR_104047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036526.mm.1 |
TC1100003909.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036529.mm.1 |
NR_104047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036529.mm.1 |
TC1100003909.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036530.mm.1 |
NR_104047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036530.mm.1 |
TC1100003909.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1430740_at |
- |
- |
Mouse Expression Set 430 |
1430740_PM_at |
- |
- |