Assay Details
| Gene Symbol: Gm15395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC048597.1 | - | 1 - 2 | 178 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polycystic kidney disease 1 like 2 | Pkd1l2 | 1700126L06Rik | Anti-sense | Chr. 8 - 116995218 - 117082488 on NCBI Build GRCm38 | |
| Downstream | beta-carotene oxygenase 1 | Bco1 |
Bcdo, Bcdo1, Bcmo1, CMO1, Cmoi, beta-CD, betaCMOOX |
57.6kb | Sense | Chr. 8 - 117095851 - 117133720 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15395
Genomic Map