Assay Details
Gene Symbol: Slc2a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130451.3 | NP_569718.1 | 3 - 4 | 1547 | 78 | |
GenBank mRNA | AK052156.1 | - | 3 - 4 | 1514 | 78 | |
AY029579.1 | - | 3 - 4 | 1470 | 78 | ||
BC137917.1 | - | 3 - 4 | 1465 | 8860936 | 78 | |
BC137920.1 | - | 3 - 4 | 1465 | 8860939 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
carbohydrate transport
glucose transport
proton transport
hexose transmembrane transport
glucose import
transmembrane transport
cation transmembrane transport
transporter activity
sugar:proton symporter activity
glucose transmembrane transporter activity
transmembrane transporter activity
substrate-specific transmembrane transporter activity
transport
carbohydrate transport
glucose transport
proton transport
hexose transmembrane transport
glucose import
transmembrane transport
cation transmembrane transport
transporter activity
sugar:proton symporter activity
glucose transmembrane transporter activity
transmembrane transporter activity
substrate-specific transmembrane transporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824617 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824254 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824614 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824258 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824619 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824615 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824259 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824257 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824618 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824616 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824256 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382912 | AK052156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382912 | AY029579.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382912 | NM_130451.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382912 | AK052156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382912 | AY029579.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P382912 | NM_130451.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644722 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591464 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639241 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957790 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818983 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574907 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 613851 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538817 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250655 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141326 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26646 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133273 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129757 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382912 | AK052156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382912 | AY029579.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382912 | NM_130451.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382912 | AK052156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382912 | AY029579.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P382912 | NM_130451.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382912 | AK052156.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382912 | AY029579.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382912 | NM_130451.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382912 | AK052156.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382912 | AY029579.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P382912 | NM_130451.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824254 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824257 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824268 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824253 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824262 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824252 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21824259 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00551808 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129757 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04775027 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133273 | - | - |
Whole Mouse Genome G4122A | A_51_P382912 | AK052156.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P382912 | AY029579.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P382912 | NM_130451.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P382912 | AK052156.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P382912 | AY029579.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P382912 | NM_130451.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106981 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136954 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129757 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133273 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01968534 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02093844 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824258 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824259 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824617 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824257 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824618 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824256 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824254 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824616 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824619 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824615 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21824614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824298 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824334 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824372 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21824244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01426081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01419942 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824274 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01426079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01291815 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02091281 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824224 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01968534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01402568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824252 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02056982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01402569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02070547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824282 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01402562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01916989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01291814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01419938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02135704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02091280 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01402563 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02091278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01603958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02056986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21824245 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10478694 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478697 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478695 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478693 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478696 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419434_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449477_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10478692 | - | - |
Mouse Genome 430 2.0 Array | 1419434_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419434_at | - | - |
Mouse Genome 430 2.0 Array | 1449477_s_at | - | - |
Mouse Genome 430 2.0 Array | 1449477_PM_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170441_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 170441_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200020939.mm.1 | NM_130451.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200020939.mm.1 | TC0200002549.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200020941.mm.1 | NM_130451.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200020941.mm.1 | TC0200002549.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200020940.mm.1 | NM_130451.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200020940.mm.1 | TC0200002549.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200020942.mm.1 | NM_130451.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200020942.mm.1 | TC0200002549.mm.1 | MTA TC |
Mouse Expression Set 430 | 1419434_PM_at | - | - |
Mouse Expression Set 430 | 1449477_PM_s_at | - | - |
Mouse Expression Set 430 | 1449477_s_at | - | - |
Mouse Expression Set 430 | 1419434_at | - | - |
Mouse Genome 430A 2.0 Array | 1419434_at | - | - |
Mouse Genome 430A 2.0 Array | 1419434_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449477_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1449477_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449477_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419434_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419434_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449477_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 385121 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 610563 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000610563 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243491 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243491 | - | - |
mouse-6 v1.1 (Illumina) | scl19914.5_100-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 610563 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl19914.5_100-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610563 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2956726 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1243491 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2956726 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243491 | - | - |
