Assay Details
| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008679.3 | NP_032705.2 | 20 - 21 | 4099 | 59 | |
| GenBank mRNA | AF000581.1 | - | 19 - 20 | 3973 | 59 | |
| AK137517.1 | - | 9 - 10 | 1770 | 59 | ||
| AK142306.1 | - | 20 - 21 | 4004 | 59 | ||
| BC086482.1 | - | 20 - 21 | 4069 | 30104508 | 59 | |
| BC093478.1 | - | 17 - 18 | 3730 | 59 | ||
| BC141177.1 | - | 19 - 20 | 3912 | 9055789 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa3
Genomic Map