| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200021339.mm.1 |
NM_030743.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021339.mm.1 |
NM_030743.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021339.mm.1 |
TC0200002588.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200021344.mm.1 |
NM_030743.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021344.mm.1 |
NM_030743.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021344.mm.1 |
TC0200002588.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200021338.mm.1 |
NM_030743.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021338.mm.1 |
NM_030743.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021338.mm.1 |
TC0200002588.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200021346.mm.1 |
NM_030743.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021346.mm.1 |
NM_030743.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200021346.mm.1 |
TC0200002588.mm.1 |
MTA TC |