Assay Details
Gene Symbol: Herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022331.1 | NP_071726.1 | 4 - 5 | 522 | 99 | |
GenBank mRNA | AB034991.1 | - | 4 - 5 | 522 | 99 | |
AK081688.1 | - | 4 - 5 | 526 | 99 | ||
AK159500.1 | - | 4 - 5 | 528 | 99 | ||
AK165832.1 | - | 4 - 5 | 528 | 99 | ||
AK172874.1 | - | 4 - 5 | 515 | 99 | ||
AK183178.1 | - | 2 - 3 | 182 | 99 | ||
BC013523.1 | - | 4 - 5 | 514 | 4236803 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
