Assay Details
| Gene Symbol: Nudt21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026623.3 | NP_080899.1 | 4 - 5 | 619 | 142 | |
| GenBank mRNA | AK011688.2 | - | 4 - 5 | 609 | 142 | |
| AK019433.1 | - | 4 - 5 | 602 | 142 | ||
| AK020150.1 | - | 4 - 5 | 600 | 142 | ||
| AK146419.1 | - | 4 - 5 | 585 | 142 | ||
| AK147061.1 | - | 602 | 142 | |||
| AK160147.1 | - | 4 - 5 | 585 | 142 | ||
| AK208594.1 | - | 3 - 4 | 194 | 142 | ||
| BC008270.1 | - | 4 - 5 | 602 | 2811199 | 142 | |
| BC090834.1 | - | 578 | 6855233 | 142 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nudt21
Genomic Map