Assay Details
| Gene Symbol: Amfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011787.2 | NP_035917.2 | 11 - 12 | 1685 | 109 | |
| GenBank mRNA | AF124144.1 | - | 11 - 12 | 1893 | 109 | |
| AK038442.1 | - | 11 - 12 | 1653 | 109 | ||
| AK157327.1 | - | 11 - 12 | 1691 | 109 | ||
| AK170716.1 | - | 11 - 12 | 1671 | 109 | ||
| AK199083.1 | - | 2 - 3 | 197 | 109 | ||
| BC003256.1 | - | 3 - 4 | 323 | 109 | ||
| BC034538.1 | - | 11 - 12 | 1643 | 4209258 | 109 | |
| BC040338.1 | - | 11 - 12 | 1611 | 3710780 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amfr
Genomic Map