Assay Details
| Gene Symbol: Slc12a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205311.1 | NP_001192240.1 | 9 - 10 | 1203 | 59 | |
| NM_019415.2 | NP_062288.2 | 9 - 10 | 1200 | 59 | ||
| GenBank mRNA | AK052691.1 | - | 9 - 10 | 1204 | 59 | |
| AK052755.1 | - | 9 - 10 | 1202 | 59 | ||
| AK085496.1 | - | 9 - 10 | 1207 | 59 | ||
| BC038612.1 | - | 9 - 10 | 1225 | 4237274 | 59 | |
| U61085.1 | - | 9 - 10 | 1182 | 59 | ||
More Information
Set Membership: |
|

Slc12a3
Genomic Map