Assay Details
| Gene Symbol: Ddx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153065.3 | NP_694705.2 | 15 - 16 | 1806 | 87 | |
| GenBank mRNA | AK134274.1 | - | 15 - 16 | 1807 | 87 | |
| AK138442.1 | - | 15 - 16 | 1830 | 87 | ||
| AK146469.1 | - | 15 - 16 | 1834 | 87 | ||
More Information
Set Membership: |
|

Ddx27
Genomic Map