Assay Details
| Gene Symbol: Ddx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153065.3 | NP_694705.2 | 18 - 19 | 2122 | 69 | |
| GenBank mRNA | AK134274.1 | - | 18 - 19 | 2123 | 69 | |
| AK138442.1 | - | 18 - 19 | 2146 | 69 | ||
| AK146469.1 | - | 18 - 19 | 2150 | 69 | ||
| AK209187.1 | - | 3 - 4 | 276 | 69 | ||
| BC011321.1 | - | 17 - 18 | 1999 | 4007058 | 69 | |
| BC026381.1 | - | 3 - 4 | 311 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx27
Genomic Map