Assay Details
Gene Symbol: Dhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144831.2 | NP_659080.2 | 20 - 21 | 3225 | 77 | |
GenBank mRNA | AK142226.1 | - | 20 - 21 | 3159 | 77 | |
AK160848.1 | - | 1166 | 77 | |||
AK170616.1 | - | 20 - 21 | 3158 | 77 | ||
AK172600.1 | - | 20 - 21 | 3173 | 77 | ||
AK220485.1 | - | 20 - 21 | 3202 | 77 | ||
BC022656.1 | - | 3 - 4 | 430 | 77 | ||
BC065064.1 | - | 20 - 21 | 3154 | 77 | ||
BC132445.1 | - | 20 - 21 | 3209 | 40130722 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
