Assay Details
| Gene Symbol: Tgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009373.3 | NP_033399.1 | 12 - 13 | 2006 | 67 | |
| GenBank mRNA | AF076928.1 | - | 11 - 12 | 1921 | 67 | |
| AF114266.1 | - | 12 - 13 | 1971 | 67 | ||
| AK052912.1 | - | 12 - 13 | 1981 | 67 | ||
| AK080224.1 | - | 12 - 13 | 2006 | 67 | ||
| AK080593.1 | - | 12 - 13 | 1981 | 67 | ||
| AK086001.1 | - | 12 - 13 | 1983 | 67 | ||
| AK089481.1 | - | 12 - 13 | 2009 | 67 | ||
| AK143712.1 | - | 12 - 13 | 2042 | 67 | ||
| AK151776.1 | - | 12 - 13 | 2007 | 67 | ||
| AK152152.1 | - | 12 - 13 | 1981 | 67 | ||
| AK152627.1 | - | 12 - 13 | 2007 | 67 | ||
| AK159255.1 | - | 12 - 13 | 1981 | 67 | ||
| AK166302.1 | - | 12 - 13 | 1981 | 67 | ||
| AK168990.1 | - | 12 - 13 | 1982 | 67 | ||
| AK169356.1 | - | 12 - 13 | 2009 | 67 | ||
| BC016492.1 | - | 12 - 13 | 1947 | 3256943 | 67 | |
| M55154.1 | - | 12 - 13 | 1940 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgm2
Genomic Map