Assay Details
| Gene Symbol: Tgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009373.3 | NP_033399.1 | 7 - 8 | 1086 | 76 | |
| GenBank mRNA | AF076928.1 | - | 6 - 7 | 1001 | 76 | |
| AF114266.1 | - | 7 - 8 | 1051 | 76 | ||
| AK052912.1 | - | 7 - 8 | 1061 | 76 | ||
| AK080224.1 | - | 7 - 8 | 1086 | 76 | ||
| AK080593.1 | - | 7 - 8 | 1061 | 76 | ||
| AK089481.1 | - | 7 - 8 | 1089 | 76 | ||
| AK143712.1 | - | 7 - 8 | 1122 | 76 | ||
| AK151776.1 | - | 7 - 8 | 1087 | 76 | ||
| AK152152.1 | - | 7 - 8 | 1061 | 76 | ||
| AK152627.1 | - | 7 - 8 | 1087 | 76 | ||
| AK159255.1 | - | 7 - 8 | 1061 | 76 | ||
| AK166302.1 | - | 7 - 8 | 1061 | 76 | ||
| AK168990.1 | - | 7 - 8 | 1062 | 76 | ||
| AK169356.1 | - | 7 - 8 | 1089 | 76 | ||
| BC016492.1 | - | 7 - 8 | 1027 | 3256943 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgm2
Genomic Map