Assay Details
| Gene Symbol: Dynlrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025947.3 | NP_080223.2 | 1 - 2 | 64 | ||
| GenBank mRNA | AK166685.1 | - | 1 - 2 | 64 | ||
| BC099423.1 | - | 1 - 2 | 30933448 | 64 | ||
| BC125610.1 | - | 1 - 2 | 40130125 | 64 | ||
| BC125648.1 | - | 1 - 2 | 40130163 | 64 | ||
| CO041603.1 | - | 3 - 4 | 619 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK166685.1, BC099423.1, BC125610.1, BC125648.1, NM_025947.3 transcripts.
Set Membership: |
|

Dynlrb1
Genomic Map