Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900026713.mm.1 |
NM_001164677.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026713.mm.1 |
NM_001164677.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026713.mm.1 |
NM_001164678.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026713.mm.1 |
NM_001164678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026713.mm.1 |
NM_011052.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026713.mm.1 |
NM_011052.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026713.mm.1 |
TC0900003160.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900026714.mm.1 |
NM_001164677.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026714.mm.1 |
NM_001164677.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026714.mm.1 |
NM_001164678.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026714.mm.1 |
NM_001164678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026714.mm.1 |
NM_011052.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026714.mm.1 |
NM_011052.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026714.mm.1 |
TC0900003160.mm.1 |
MTA TC |