Assay Details
| Gene Symbol: Ccdc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012309.2 | NP_001012309.1 | 3 - 4 | 204 | 101 | |
| GenBank mRNA | AK041865.1 | - | 3 - 4 | 190 | 101 | |
| AK046155.1 | - | 3 - 4 | 190 | 101 | ||
| AK078762.1 | - | 3 - 4 | 190 | 101 | ||
| AK078945.1 | - | 3 - 4 | 191 | 101 | ||
| AK162143.1 | - | 3 - 4 | 191 | 101 | ||
| AK168573.1 | - | 3 - 4 | 190 | 101 | ||
| BC016601.1 | - | 3 - 4 | 179 | 101 | ||
| BC089560.1 | - | 3 - 4 | 237 | 30300075 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc55
Genomic Map