Assay Details
Gene Symbol: Slc6a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF013604.1 | - | 10 - 11 | 1546 | 71 | |
BC108978.1 | - | 11 - 12 | 1715 | 40047534 | 71 | |
BC108979.1 | - | 11 - 12 | 1715 | 40047537 | 71 | |
X66119.1 | - | 10 - 11 | 1327 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
