Assay Details
| Gene Symbol: Lrrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028838.2 | NP_083114.2 | 3 - 4 | 491 | 60 | |
| GenBank mRNA | AJ428068.1 | - | 3 - 4 | 490 | 60 | |
| AK010252.1 | - | 3 - 4 | 490 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc2
Genomic Map