Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900026406.mm.1 |
NM_001083935.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026406.mm.1 |
NM_001083935.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026406.mm.1 |
NM_001083936.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026406.mm.1 |
NM_001083936.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026406.mm.1 |
NM_011199.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026406.mm.1 |
NM_011199.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900026406.mm.1 |
TC0900003116.mm.1 |
MTA TC |