Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800023875.mm.1 |
NM_009824.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023875.mm.1 |
NM_009824.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023875.mm.1 |
TC0800003080.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023874.mm.1 |
NM_009824.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023874.mm.1 |
NM_009824.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023874.mm.1 |
TC0800003080.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023873.mm.1 |
NM_009824.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023873.mm.1 |
NM_009824.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023873.mm.1 |
TC0800003080.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023868.mm.1 |
NM_001109873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023868.mm.1 |
NM_001109873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023868.mm.1 |
NM_009824.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023868.mm.1 |
NM_009824.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023868.mm.1 |
NM_177289.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023868.mm.1 |
NM_177289.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023868.mm.1 |
TC0800003080.mm.1 |
MTA TC |