Assay Details
Gene Symbol: Scimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045526.2 | NP_001038991.1 | 4 - 5 | 346 | 87 | |
GenBank mRNA | AK138818.1 | - | 4 - 5 | 347 | 87 | |
BC147177.1 | - | 4 - 5 | 344 | 9056232 | 87 | |
BC147178.1 | - | 4 - 5 | 344 | 9056215 | 87 | |
BC172005.1 | - | 4 - 5 | 340 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963920 | NM_001045526.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963920 | AK138818.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604739 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338673 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822721 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662416 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423398 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668832 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171495 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662545 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121271 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 800191 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851219 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953845 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953757 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95227 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88070 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156880 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147462 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963920 | NM_001045526.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963920 | AK138818.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02357983 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147462 | - | - |
Whole Mouse Genome G4122A | A_52_P638457 | NM_001045526.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P638457 | AK138818.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P638457 | BC147177.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P638457 | BC147178.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P638457 | BC172005.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27902733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09295944 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09676458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902812 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902834 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09295948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09013843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09286991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902841 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09295942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09695101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09584237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09520328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902801 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902837 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902846 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902826 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09571452 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09056028 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09571450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09574841 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902802 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09286992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09285850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902859 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09571451 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902844 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902694 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09437300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27902701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09013842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09295936 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10387986 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387988 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387989 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387987 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387991 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10387990 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10387985 | - | - |
Affymetrix Mouse Clariom S Array | PSR1100028782.mm.1 | NM_001045526.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028782.mm.1 | NM_001045526.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028782.mm.1 | TC1100003115.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100028783.mm.1 | NM_001045526.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028783.mm.1 | NM_001045526.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100028783.mm.1 | TC1100003115.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 472042 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2534201 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2847437 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2847437 | - | - |
