Assay Details
| Gene Symbol: Rgs9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165934.1 | NP_001159406.1 | 3 - 4 | 395 | 128 | |
| NM_011268.2 | NP_035398.2 | 3 - 4 | 395 | 128 | ||
| GenBank mRNA | AF011358.1 | - | 3 - 4 | 280 | 128 | |
| AF125046.1 | - | 3 - 4 | 351 | 128 | ||
| AK085443.1 | - | 3 - 4 | 321 | 128 | ||
| AK138159.1 | - | 3 - 4 | 305 | 128 | ||
| AK164961.1 | - | 3 - 4 | 349 | 128 | ||
| AK166518.1 | - | 239 | 128 | |||
| BC129899.1 | - | 3 - 4 | 261 | 40111531 | 128 | |
More Information
Set Membership: |
|

Rgs9
Genomic Map